Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-4'-hydroxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. C12391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12391G 1 g
C12395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12391G Supplier TCI America Supplier No. C12391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28034-99-3
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.653
MDL Number MFCD00143298
InChI Key ICVFJPSNAUMFCW-UHFFFAOYSA-N
Synonym 4-chloro-4'-hydroxybiphenyl, 4-chloro-4'-biphenylol, 4'-chlorobiphenyl-4-ol, 4-4-chlorophenyl phenol, 4-chloro-4-hydroxybiphenyl, 4-biphenylol, 4'-chloro, unii-in91367n9r, 1,1'-biphenyl-4-ol, 4'-chloro, 4'-chloro-4-biphenylol, 4'-chloro-1,1'-biphenyl-4-ol
PubChem CID 91589
ChEBI CHEBI:34394
IUPAC Name 4-(4-chlorophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)O

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 204.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-4′-hydroxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.