Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chloro-5-(3,4-dimethylphenyl)thieno[2,3-d]pyrimidine

Catalog No. OTV000580 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000580 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000580 Supplier Sigma Aldrich Supplier No. OTV000580250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylphosphoramic dichloride can be used as a reagent:
  • For the activation of carboxylic acids
  • To prepare functionalized α-lactams by reacting with acetic acids and imines
  • To synthesize phosphoramidic esters by reacting with alcohols in the presence of a base catalyst
  • To prepare [1,2,4]triazolo[4,3-c][1,3,5,2]triazaphosphinine-5-oxide derivatives by reacting with n-alkyl/aryl-nα-(4h-1,2,4-triazol-3-yl) amidines in the presence of triethylamine

Specifications

CAS 379241-56-2
Molecular Formula C14H11ClN2S
Linear Formula C14H11ClN2S
MDL Number MFCD03001383
Quantity 10 mL
Molecular Weight (g/mol) 274.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.