Learn More
Sigma Aldrich 4-Chloro-5-(4-ethoxyphenyl)-2,6-dimethylthieno[2,3-d]pyrimidine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Farnesal has been reported as specific lipid substrate for aldo-keto reductase 1b10 (akr1b10)
- The incubation of farnesol with the protoplast of botryococcus braunii b race strain leads to the formation of farnesal and 3-hydroxy-2,3-dihydrofarnesal.farnesal was used in the synthesis of sesquiterpene nanaimoal
Specifications
Specifications
| Molecular Formula | C16H15ClN2OS |
| Linear Formula | C16H15ClN2OS |
| Quantity | 1 mL |
| MDL Number | MFCD10009768 |
| Molecular Weight (g/mol) | 318.82 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.