Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chloro-5-(4-fluorophenyl)-2,6-dimethylthieno[2,3-d]pyrimidine

Catalog No. OTV000397 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000397 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000397 Supplier Sigma Aldrich Supplier No. OTV000397250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-(2-hydroxyethyl)-2-imidazolidinone has been identified as monoethanolamine (mea) degradation product in imc chemicals facility in trona, ca plant performing co2 capture from flue gas
  • It was also identified from the mea reclaimer from a co2 capture facility product
  • It was also identified as the most stable thermal degradation product of mea.reagent for synthesis of hiv-1 integrase inhibitorsreactant for synthesis of: mdm2-p53 protein-protein interaction inhibitors amino acid-derived heterocycles dopamine d-2 and serotonin 5-ht2 antagonists

Specifications

Molecular Formula C14H10ClFN2S
Linear Formula C14H10ClFN2S
MDL Number MFCD10009780
Quantity 250 mL
Molecular Weight (g/mol) 292.76
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.