Learn More
Sigma Aldrich 4-Chloro-5-(4-isopropylphenyl)thieno[2,3-d]pyrimidine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-(trifluoroacetyl)imidazole (n-(trifluroroacetyl)imidazole) is an effective reagent for specific trifluoroacetylation of an aminomethyl side chain of certain nucleosides.1-(trifluoroacetyl)imidazole may be employed as derivatization reagent for the detection of mustard gas degradation products, thiodiglycol and thiodiglycol sulfoxide using gas chromatography-tandem mass spectrometry
- It may be used as reagent for the trifluoroacetylation of amines
- It was used for derivatization of ethanolamine phospholipid-alkenal michael adducts for GC-ms analysis
- It was used in the determination of quinolinic acid, a kynurenine metabolite, by GC/ms
Specifications
Specifications
| CAS | 610274-03-8 |
| Molecular Formula | C15H13ClN2S |
| Linear Formula | C15H13ClN2S |
| MDL Number | MFCD06253954 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 288.8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.