Learn More
Medchemexpress LLC 4-chloro-5-cyano-N-[2-(diethylamino)ethyl]-2-methoxybenzamide | 104391-26-6 | 99.7% | 346.25 | C15H21Cl2N3O2 | 100 MG
SDP
Catalog No. 5000214855
Encompass_Preferred
CGP 25454A is a selective presynaptic dopamine autoreceptor antagonist used in neuroscience research to increase extracellular dopamine and acetylcholine levels. Supplied as a white to off-white solid, it is suitable for in vitro and in vivo studies; certificate of analysis and safety data sheet are available.
Catalog No. 50-002-14855 Supplier Medchemexpress LLC Supplier No. HY100454100MG
CGP 25454A is a selective presynaptic dopamine autoreceptor antagonist used in neuroscience research to increase extracellular dopamine and acetylcholine levels. Supplied as a white to off-white solid, it is suitable for in vitro and in vivo studies; certificate of analysis and safety data sheet are available.