Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™
SDP

Catalog No. C30081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30081G 1 g
C3008200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30081G Supplier TCI America Supplier No. C30081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116941-51-6
Molecular Formula C24H17Cl
Molecular Weight (g/mol) 340.85
InChI Key FZOIFXXPBLTVIG-UHFFFAOYSA-N
Synonym 5′C-(4-Chlorophenyl)-1,1′C:3′C,1′C′C-terphenyl
PubChem CID 11110599
IUPAC Name 1-(4-chlorophenyl)-3,5-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4

Specifications

Melting Point 143°C
Color White
Quantity 1 g
Formula Weight 340.85
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-5′-phenyl-1,1′:3′,1′′-terphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.