Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-methylquinoline, 97%, Thermo Scientific Chemicals

Catalog No. AAH5989003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5989003 Supplier Thermo Scientific Chemicals Supplier No. H5989003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18436-71-0 | Molecular Formula: C10H8ClN | Molecular Weight: 177.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18436-71-0
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD02684204
InChI Key HZWWPOQFLMUYOX-UHFFFAOYSA-N
Synonym quinoline, 4-chloro-6-methyl, 4-chloro-6-methyl-quinoline, acmc-1c6ji, ksc497i2j, 4-chloranyl-6-methyl-quinoline
PubChem CID 824624
IUPAC Name 4-chloro-6-methylquinoline
SMILES CC1=CC2=C(C=CN=C2C=C1)Cl

Specifications

Quantity 1 g
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-methylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.