Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-chloro-7-fluoropyrrolo[1,2-a]quinoxaline | 136773-69-8 | MFCD09835554 | 98.2% | 10 MG
SDP

Catalog No. 5000297618
Encompass_Preferred

4-Chloro-7-fluoropyrrolo[1,2-a]quinoxaline is a small-molecule drug intermediate used in medicinal chemistry for the synthesis of biologically active compounds. It is supplied as a solid powder in research-scale quantities and has reported analytical data supporting its use in organic synthesis and method development.

  • High purity suitable for synthetic use (reported 98.2%).
  • Molecular weight 220.63 g/mol.
  • Chemical formula C11H6ClFN2.
  • Solid powder physical form for handling and storage.
  • Intended for use as a synthetic intermediate in medicinal chemistry.
  • Available in milligram-scale packages for research applications.

Catalog No. 50-002-97618 Supplier Medchemexpress LLC Supplier No. HY2369510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Chloro-7-fluoropyrrolo[1,2-a]quinoxaline is a small-molecule drug intermediate used in medicinal chemistry for the synthesis of biologically active compounds. It is supplied as a solid powder in research-scale quantities and has reported analytical data supporting its use in organic synthesis and method development.

  • High purity suitable for synthetic use (reported 98.2%).
  • Molecular weight 220.63 g/mol.
  • Chemical formula C11H6ClFN2.
  • Solid powder physical form for handling and storage.
  • Intended for use as a synthetic intermediate in medicinal chemistry.
  • Available in milligram-scale packages for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.