Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine | 3680-69-1 | 99.82% | 153.57 | 25 G
SDP

Catalog No. 5000402184
Encompass_Preferred

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine is a key intermediate in the preparation of active pharmaceutical ingredients.

  • Purity: 99.82%
  • Data sheet, COA, SDS, and handling instructions are available
  • For research use only; not sold to patients
  • Available in quantities such as 25 g and 50 g
  • Shipping is at room temperature in the continental US; may vary elsewhere
  • Storage recommendations: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months, or -20°C for 1 month (sealed storage, away from moisture)

Catalog No. 50-004-02184 Supplier Medchemexpress LLC Supplier No. 25GHY40351
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine is a key intermediate in the preparation of active pharmaceutical ingredients.

  • Purity: 99.82%
  • Data sheet, COA, SDS, and handling instructions are available
  • For research use only; not sold to patients
  • Available in quantities such as 25 g and 50 g
  • Shipping is at room temperature in the continental US; may vary elsewhere
  • Storage recommendations: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months, or -20°C for 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.