Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-chloro-8-(2,4-dichlorophenyl)-2,7-dimethylpyrazolo[1,5-a]triazine | 202578-52-7 | 98.7% | 10 MG
SDP

Catalog No. 5000231413
Encompass_Preferred

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used as a research compound in preclinical studies of anxiety, depression, and fear-memory. It shows in vivo activity in rodent behavioral models and is supplied as a solid powder for laboratory use.

  • Selective CRHR1 antagonist for preclinical research.
  • Demonstrated activity in rodent behavioral and fear-memory models.
  • Molecular weight 410.30 g/mol; formula C18H21Cl2N5O2.
  • Supplied as a solid powder, commonly in 10 mg packages.
  • Typical purity approximately 98.7% (batch-specific).
  • Storage: powder -20°C for long-term; in solvent -80°C for extended storage.

Catalog No. 50-002-31413 Supplier Medchemexpress LLC Supplier No. HY1213110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used as a research compound in preclinical studies of anxiety, depression, and fear-memory. It shows in vivo activity in rodent behavioral models and is supplied as a solid powder for laboratory use.

  • Selective CRHR1 antagonist for preclinical research.
  • Demonstrated activity in rodent behavioral and fear-memory models.
  • Molecular weight 410.30 g/mol; formula C18H21Cl2N5O2.
  • Supplied as a solid powder, commonly in 10 mg packages.
  • Typical purity approximately 98.7% (batch-specific).
  • Storage: powder -20°C for long-term; in solvent -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.