Learn More
AdipoGen 4-Chloro-2,1,3-benzoxadiazole (10 g)
SDP
Supplier:  AdipoGen CDXC0577G010
Encompass_Preferred
Chemical. CAS: 7116-16-7. Formula: C6H3CIN2O. MW: 154.55. Building block used for the synthesis of fluorescent probes.
Catalog No. 50-205-4335