Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-D-mandelic Acid 98.0+%, TCI America™
SDP

Catalog No. C30061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30061G 1 g
C30065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30061G Supplier TCI America Supplier No. C30061G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32189-36-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798430
InChI Key BWSFWXSSALIZAU-SSDOTTSWSA-N
Synonym (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid
PubChem CID 785085
IUPAC Name (2R)-2-(4-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)Cl

Specifications

Melting Point 121°C
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-Chloro-D-mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.