Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-D-phenylalanine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C20161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20161G Supplier TCI America Supplier No. C20161G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 147065-05-2
Molecular Formula C9H11Cl2NO2
Molecular Weight (g/mol) 236.092
InChI Key PFOCEDBJFKVRHU-DDWIOCJRSA-N
Synonym (R)-2-Amino-3-(4-chlorophenyl)propionic Acid Hydrochloride, H-D-Phe(4-Cl)-OH.HCl
PubChem CID 45357975
IUPAC Name (2R)-2-amino-3-(4-chlorophenyl)propanoic acid;hydrochloride
SMILES C1=CC(=CC=C1CC(C(=O)O)N)Cl.Cl

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 1 g
Formula Weight 236.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-D-phenylalanine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.