Learn More
Selleck Chemical LLC 4-Chloro-DL-phenylalanine S4586-100mg
SDP
Catalog No. 503111555
Encompass_Preferred
4-Chloro-DL-phenylalanine (Fenclonine PCPA CP-10188) is a selective and irreversible inhibitor of tryptophan hydroxylase a rate-limiting enzyme in the biosynthesis of serotonin (5-HYDROXYTRYPTAMINE) 4-Chloro-DL-phenylalanine acts pharmacologically to deplete endogenous levels of serotonin
Catalog No. 50-311-1555 Supplier Selleck Chemical LLC Supplier No. S4586100MG
4-Chloro-DL-phenylalanine (Fenclonine PCPA CP-10188) is a selective and irreversible inhibitor of tryptophan hydroxylase a rate-limiting enzyme in the biosynthesis of serotonin (5-HYDROXYTRYPTAMINE) 4-Chloro-DL-phenylalanine acts pharmacologically to deplete endogenous levels of serotonin