Learn More
Medchemexpress LLC 4-Chloro-DL-phenylalanine methyl ester hydrochloride | 14173-40-1 | MFCD00012530 | 99.9% | 250.12 | C10H13Cl2NO2 | 5 G
SDP
Catalog No. 5000247949
Encompass_Preferred
PCPA methyl ester hydrochloride is the hydrochloride salt of 4-chloro-DL-phenylalanine methyl ester provided for research use. It acts as a tryptophan hydroxylase inhibitor and is used in neurochemical and pharmacological studies to reduce central serotonin (5-HT) levels. The compound is supplied as a high-purity solid suitable for biochemical and in vivo experiments.
  • High purity suitable for research applications.
  • Inhibits tryptophan hydroxylase to reduce serotonin synthesis.
  • Crosses the blood-brain barrier for central nervous system studies.
  • Provided as a stable hydrochloride salt for reproducible dosing.
  • Available in multiple lab-scale pack sizes for experimental flexibility.
Catalog No. 50-002-47949 Supplier Medchemexpress LLC Supplier No. HY1014565G
PCPA methyl ester hydrochloride is the hydrochloride salt of 4-chloro-DL-phenylalanine methyl ester provided for research use. It acts as a tryptophan hydroxylase inhibitor and is used in neurochemical and pharmacological studies to reduce central serotonin (5-HT) levels. The compound is supplied as a high-purity solid suitable for biochemical and in vivo experiments.
  • High purity suitable for research applications.
  • Inhibits tryptophan hydroxylase to reduce serotonin synthesis.
  • Crosses the blood-brain barrier for central nervous system studies.
  • Provided as a stable hydrochloride salt for reproducible dosing.
  • Available in multiple lab-scale pack sizes for experimental flexibility.