Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide 98.0+%, TCI America™
SDP

Catalog No. C23091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23091G 1 g
C23095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23091G Supplier TCI America Supplier No. C23091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41859-57-8
Molecular Formula C15H14ClNO2
Molecular Weight (g/mol) 275.73
MDL Number MFCD00218513
InChI Key ZTLWJYCDAXUIBK-UHFFFAOYSA-N
Synonym n-4-chlorobenzoyl-tyramine, 4-chloro-n-4-hydroxyphenethyl benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, n-4-chlorobenzoyl tyramine, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, bez-da
PubChem CID 198234
IUPAC Name 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
SMILES OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 174°C
Quantity 1 g
Formula Weight 275.73
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
RTECSNumber CV2450481
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.