Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-n-hexylbutyramide, 97%, Thermo Scientific™

Catalog No. AAH59434MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59434MD 250 mg
AAH5943403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59434MD Supplier Thermo Scientific Chemicals Supplier No. H59434MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543715-14-6
Molecular Formula C10H20ClNO
Molecular Weight (g/mol) 205.726
MDL Number MFCD03375372
InChI Key RAEKTBRIUBNXFJ-UHFFFAOYSA-N
Synonym 4-chloro-n-n-hexylbutyramide
PubChem CID 4097406
IUPAC Name 4-chloro-N-hexylbutanamide
SMILES CCCCCCNC(=O)CCCCl

Specifications

Quantity 250 mg
Formula Weight 205.72
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-n-hexylbutyramide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.