Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroanisole, 99%

Catalog No. AC170151000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170151000 100 g Glass bottle
AC170150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170151000 Supplier Thermo Scientific Chemicals Supplier No. 170151000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 623-12-1 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-12-1
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
InChI Key YRGAYAGBVIXNAQ-UHFFFAOYSA-N
Synonym 4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
PubChem CID 12167
IUPAC Name 1-chloro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -18.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 198.0°C to 202.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5340 to 1.5360
Linear Formula ClC6H4OCH3
Quantity 100 g
Beilstein 06,186
Specific Gravity 1.16
Solubility Information Solubility in water: immiscible.
Formula Weight 142.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chloroanisole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 210-772-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.