Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-chlorobenzene-1,3-diol | 95-88-5 | MFCD00002273 | 98.6% | 144.56 g/mol | C6H5ClO2 | 500 G
SDP

Catalog No. 5000300055
Encompass_Preferred

4-Chlorobenzene-1,3-diol (4-chlororesorcinol; CAS 95-88-5) is an organic biochemical reagent used in life-science research and analytical applications. Supplied as a solid, it is commonly provided at high purity for synthetic chemistry, analytical standards, and biochemical studies.

  • Chemical identity: 4-chlorobenzene-1,3-diol (CAS 95-88-5).
  • Molecular formula: C6H5ClO2; molecular weight: 144.56 g/mol.
  • High purity suitable for research use (reported ~98.6%).
  • Available as a 500 g pack for bulk laboratory use.
  • Solubility: soluble in DMSO for stock solution preparation.
  • Applications: analytical standards, synthetic building block, and biochemical research.

Catalog No. 50-003-00055 Supplier Medchemexpress LLC Supplier No. HYW015658500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Chlorobenzene-1,3-diol (4-chlororesorcinol; CAS 95-88-5) is an organic biochemical reagent used in life-science research and analytical applications. Supplied as a solid, it is commonly provided at high purity for synthetic chemistry, analytical standards, and biochemical studies.

  • Chemical identity: 4-chlorobenzene-1,3-diol (CAS 95-88-5).
  • Molecular formula: C6H5ClO2; molecular weight: 144.56 g/mol.
  • High purity suitable for research use (reported ~98.6%).
  • Available as a 500 g pack for bulk laboratory use.
  • Solubility: soluble in DMSO for stock solution preparation.
  • Applications: analytical standards, synthetic building block, and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.