Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzotrichloride 98.0+%, TCI America™
SDP

Catalog No. T007225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T007225G Supplier TCI America Supplier No. T007225G

missing translation for 'validMainframeMsg'

Specifications

Melting Point 6°C
CAS 5216-25-1
Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Molecular Formula C7H4Cl4
MDL Number MFCD00000790
Synonym 4-chlorobenzotrichloride, 1-chloro-4-trichloromethyl benzene, p-chlorobenzotrichloride, benzene, 1-chloro-4-trichloromethyl, p-chlorophenyltrichloromethane, p-trichloromethylchlorobenzene, alpha,alpha,alpha,4-tetrachlorotoluene, unii-1up9gju33s, ccris 8902, p,alpha,alpha,alpha-tetrachlorotoluene
Boiling Point 245°C
InChI Key LVZPKYYPPLUECL-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl
IUPAC Name 1-chloro-4-(trichloromethyl)benzene
Molecular Weight (g/mol) 229.909
PubChem CID 21277
Formula Weight 229.91
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chlorobenzotrichloride
Show More Show Less
EINECSNumber (3)-0089&(3)-0093
RTECSNumber XT8580000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.