Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzyl Mercaptan 98%, Thermo Scientific™

Catalog No. AC108821000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC108821000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC108821000 Supplier Thermo Scientific Chemicals Supplier No. 108821000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6258-66-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H7ClS
Linear Formula ClC6H4CH2SH
MDL Number MFCD00004870
Beilstein 06, 466
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
SMILES SCC1=CC=C(Cl)C=C1
IUPAC Name (4-chlorophenyl)methanethiol
Molecular Weight (g/mol) 158.64
PubChem CID 80409
Formula Weight 158.65
Percent Purity ≥97.5%
Chemical Name or Material 4-Chlorobenzyl mercaptan, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-395-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.