Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzyl Mercaptan 98.0+%, TCI America™
SDP

Catalog No. C065825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065825G 25 g
C0658500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C065825G Supplier TCI America Supplier No. C065825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6258-66-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00004870
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
PubChem CID 80409
IUPAC Name (4-chlorophenyl)methanethiol
SMILES SCC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Boiling Point 103°C
Formula Weight 158.64
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chlorobenzyl Mercaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.