Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chlorobiphenyl-4-sulfonyl chloride, 96%

Catalog No. AAH3418209
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3418209 10 g
AAH3418203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3418209 Supplier Thermo Scientific Chemicals Supplier No. H3418209
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20443-74-7 | C12H8Cl2O2S | 287.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20443-74-7
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.154
MDL Number MFCD01631918
InChI Key NWYUSJMIHFIMTA-UHFFFAOYSA-N
Synonym 4'-chlorobiphenyl-4-sulfonyl chloride, 4-4-chlorophenyl benzenesulfonyl chloride, 4'-chloro-biphenyl-4-sulfonyl chloride, 4'-chloro-1,1'-biphenyl-4-sulfonyl chloride, 4'-chloro-4-biphenylylsulphonyl chloride, 4'-chloro 1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-chloro, 4'-chlorobiphenyl4-sulfonyl chloride, 4-chlorobiphenyl-4'-sulfonyl chloride, 4'-chloro biphenyl-4-sulfonyl chloride
PubChem CID 2794745
IUPAC Name 4-(4-chlorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)Cl

Specifications

Melting Point 107°C to 108°C
Quantity 10 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 287.17
Percent Purity 96%
Chemical Name or Material 4'-Chlorobiphenyl-4-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.