Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyrophenone, 95%, Thermo Scientific™

Catalog No. p-7044432
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0638306 5 g
AAL0638314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0638306 Supplier Thermo Scientific Chemicals Supplier No. L0638306
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-52-6
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00001005
InChI Key GHEFQKHLHFXSBR-UHFFFAOYSA-N
Synonym 4-chlorobutyrophenone, gamma-chlorobutyrophenone, 4-chloro-1-oxo-1-phenylbutane, .gamma.-chlorobutyrophenone, 1-butanone,4-chloro-1-phenyl, chlorpropylphenylketon, 4-chlorobutyrylbenzene, 4-chlorobutyro-phenone, 3-benzoylpropyl chloride, .omega.-chlorobutyrophenone
PubChem CID 253533
IUPAC Name 4-chloro-1-phenylbutan-1-one
SMILES ClCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 19°C to 20°C
Density 1.137
Boiling Point 138°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 5 g
Beilstein 878974
Formula Weight 182.65
Percent Purity 95%
Chemical Name or Material 4-Chlorobutyrophenone
EINECSNumber 213-362-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.