Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chlorobutyrophenone 98.0+%, TCI America™
SDP

Catalog No. C154725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C154725G Supplier TCI America Supplier No. C154725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4981-63-9
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00013647
InChI Key XLCJPQYALLFIPW-UHFFFAOYSA-N
Synonym 4'-chlorobutyrophenone, p-chlorobutyrophenone, 1-4-chlorophenyl butan-1-one, 1-butanone, 1-4-chlorophenyl, butyrophenone, 4'-chloro, 4-chlorophenyl propyl ketone, acmc-1al89, xlcjpqyallfipw-uhfffaoysa, 1-4-chlorophenyl-1-butanone #
PubChem CID 70321
IUPAC Name 1-(4-chlorophenyl)butan-1-one
SMILES CCCC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 254°C
Quantity 25 g
Formula Weight 182.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chlorobutyrophenone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.