Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyryl Chloride, Spectrum™ Chemical
SDP

Catalog No. 18620424 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-424 500 mL
18-620-423 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-424 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C2226500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4635-59-0

Specifications

Boiling Point 173.5°C/344°F
Quantity 500 mL
Molecular Weight (g/mol) 141
Percent Purity 98%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyryl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.