Learn More
Medchemexpress LLC 4-chlorocinnamic acid | 1615-02-7 | MFCD00004396 | 100.0% | 182.61 | C9H7ClO2 | 50g
SDP
Catalog No. 5000267878
Encompass_Preferred
4-Chlorocinnamic acid is a derivative of Cinnamic acid (HY-N0610A) 4-Chlorocinnamic acid inhibits the monophenolase and diphenolase activities of mushroom tyrosinase with IC50 values of 0 477 mM and 0 229 mM respectively 4-Chlorocinnamic acid inhibits the mycelial growth of Colletotrichum gloeosporioides 4-Chlorocinnamic acid exhibits anticonvulsant activity against acute epilepsy induced by pentylenetetrazol[1][2][3][4]
Catalog No. 50-002-67878 Supplier Medchemexpress LLC Supplier No. HYY072950G
4-Chlorocinnamic acid is a derivative of Cinnamic acid (HY-N0610A) 4-Chlorocinnamic acid inhibits the monophenolase and diphenolase activities of mushroom tyrosinase with IC50 values of 0 477 mM and 0 229 mM respectively 4-Chlorocinnamic acid inhibits the mycelial growth of Colletotrichum gloeosporioides 4-Chlorocinnamic acid exhibits anticonvulsant activity against acute epilepsy induced by pentylenetetrazol[1][2][3][4]