Learn More
Medchemexpress LLC 4-Chloroindole-3-acetic acid | 2519-61-1 | MFCD00216155 | 209.63 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00626710G
Encompass_Preferred
4-Chloroindole-3-acetic acid (4-Cl-IAA) is a chlorinated analogue of indole-3-acetic acid (IAA) auxin. It functions as a plant hormone and is naturally found in the seeds of *Medicago truncatula*, *Melilotus indicus*, and *Trifolium*.
  • Functions as a plant hormone
  • Chlorinated analogue of indole-3-acetic acid (IAA) auxin
  • Found in seeds of *Medicago truncatula*, *Melilotus indicus*, and *Trifolium*
  • Appears as a white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Solid can be stored at room temperature for 3 years
  • Solutions can be stored at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-48582