Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroisatin 97.0+%, TCI America™
SDP

Catalog No. C18705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18705G 5 g
C187025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18705G Supplier TCI America Supplier No. C18705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6344-05-4
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.58
InChI Key HSYFISNDMZKGRS-UHFFFAOYSA-N
Synonym 4-Chloroindole-2,3-dione, 4-Chloro-2,3-indolinedione
PubChem CID 96047
IUPAC Name 4-chloro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C(=O)N2

Specifications

Melting Point 258°C
Quantity 5 g
Formula Weight 181.58
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloroisatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.