Learn More
Accela Chembio Inc 4-chloroisobenzofuran-1 | 3-dione | 25g | 117-21-5 | MFCD00023107 | 96% | Shelf Life: 1800 Days | Moisture Sensitive
Catalog No. 50353101
Encompass_Preferred
4-chloroisobenzofuran-1 | 3-dione | 25g | 117-21-5 | MFCD00023107 | 96% | Shelf Life: 1800 Days | Moisture Sensitive
Catalog No. 50-353-101 Supplier Accela Chembio Inc Supplier No. SY00100125G
4-chloroisobenzofuran-1 | 3-dione | 25g | 117-21-5 | MFCD00023107 | 96% | Shelf Life: 1800 Days | Moisture Sensitive