Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Chloromethyl)-2-isopropylthiazole

Catalog No. SYX003971G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003971G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003971G Supplier Sigma Aldrich Supplier No. SYX003971G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dichlorotoluene is added as growth supplement in culture medium of ralstonia sp
  • Strain ps12.3,4-dichlorotoluene was used in determination of enthalpy of formation for molecular complexes of i2 with chloromethylbenzene molecules in ccl4

Specifications

CAS 40516-57-2
Molecular Formula C7H10ClNS
Linear Formula C7H10ClNS
MDL Number MFCD06660126
Quantity 100 g
Molecular Weight (g/mol) 175.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.