Learn More
Medchemexpress LLC 4-(Chloromethyl)-7-hydroxycoumarin | 25392-41-0 | 98.0% | C10H7ClO3 | 100 MG
SDP
Catalog No. 5000434038
Encompass_Preferred
4-(Chloromethyl)-7-hydroxycoumarin is a hydroxycoumarin derivative that exhibits potent antioxidant effects and high hydroxyl radical-scavenging properties. It contains a methyl group and a chlorine group within its heterocyclic ring. This compound has been investigated as a potent inhibitor of the zinc enzyme carbonic anhydrase, particularly the tumor-associated isoforms CA IX and XII.
  • Exhibits potent antioxidant effects
  • Possesses high hydroxyl radical-scavenging properties
  • Contains a methyl group and a chlorine group in its heterocyclic ring
  • Investigated as a potent inhibitor of zinc enzyme carbonic anhydrase
  • Shows inhibitory activity against tumor-associated isoforms CA IX and XII
Catalog No. 50-004-34038 Supplier Medchemexpress LLC Supplier No. HY76779100MG
4-(Chloromethyl)-7-hydroxycoumarin is a hydroxycoumarin derivative that exhibits potent antioxidant effects and high hydroxyl radical-scavenging properties. It contains a methyl group and a chlorine group within its heterocyclic ring. This compound has been investigated as a potent inhibitor of the zinc enzyme carbonic anhydrase, particularly the tumor-associated isoforms CA IX and XII.
  • Exhibits potent antioxidant effects
  • Possesses high hydroxyl radical-scavenging properties
  • Contains a methyl group and a chlorine group in its heterocyclic ring
  • Investigated as a potent inhibitor of zinc enzyme carbonic anhydrase
  • Shows inhibitory activity against tumor-associated isoforms CA IX and XII