Learn More
Medchemexpress LLC 4-(Chloromethyl)-7-hydroxycoumarin | 25392-41-0 | 98.0% | C10H7ClO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY76779
Encompass_Preferred
4-(Chloromethyl)-7-hydroxycoumarin is a hydroxycoumarin derivative exhibiting potent antioxidant effects and high hydroxyl radical-scavenging properties. It contains a methyl group and a chlorine group within its heterocyclic ring. This compound has been investigated for its ability to inhibit the zinc enzyme carbonic anhydrase, particularly tumor-associated isoforms CA IX and XII.
  • Exhibits potent antioxidant effect
  • Possesses high hydroxyl radical-scavenging property
  • Contains methyl and chlorine groups in the heterocyclic ring
  • Investigated as an inhibitor of zinc enzyme carbonic anhydrase
  • Targets tumor-associated isoforms CA IX and XII
  • For research use only
Catalog No. 50-003-58040