Learn More
Medchemexpress LLC 4-(Chloromethyl)-7-hydroxycoumarin | 25392-41-0 | 98.01% | C10H7ClO3 | 1 ML
SDP
Catalog No. 5000434064
Encompass_Preferred
4-(Chloromethyl)-7-hydroxycoumarin is a hydroxycoumarin derivative exhibiting potent antioxidant effects and high hydroxyl radical-scavenging properties. It contains a methyl group and a chlorine group within its heterocyclic ring and has been investigated as a potent inhibitor of zinc enzyme carbonic anhydrase isoforms CA IX and XII.
  • Potent antioxidant effect
  • High hydroxyl radical-scavenging property
  • Investigated as inhibitors of tumor-associated carbonic anhydrase isoforms CA IX and XII
Catalog No. 50-004-34064 Supplier Medchemexpress LLC Supplier No. HY7677910MM/1ML
4-(Chloromethyl)-7-hydroxycoumarin is a hydroxycoumarin derivative exhibiting potent antioxidant effects and high hydroxyl radical-scavenging properties. It contains a methyl group and a chlorine group within its heterocyclic ring and has been investigated as a potent inhibitor of zinc enzyme carbonic anhydrase isoforms CA IX and XII.
  • Potent antioxidant effect
  • High hydroxyl radical-scavenging property
  • Investigated as inhibitors of tumor-associated carbonic anhydrase isoforms CA IX and XII