Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Chloromethyl)pyridine hydrochloride

Catalog No. P4380710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P4380710G 10 g
P43807100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P4380710G Supplier Sigma Aldrich Supplier No. P4380710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-(Chloromethyl)pyridine hydrochloride

Specifications

CAS 1822-51-1
Melting Point 166°C to 173°C (lit.)
Molecular Formula C6H6ClN -+ HCl
Linear Formula C6H6ClN -+ HCl
MDL Number MFCD00012826
Quantity 10 g
Synonym 4-Picolyl chloride hydrochloride
Molecular Weight (g/mol) 164.03
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.