Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenetole 99.0+%, TCI America™
SDP

Catalog No. C023925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C023925G Supplier TCI America Supplier No. C023925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-61-7
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00058931
InChI Key IXLSVQMYQRAMEW-UHFFFAOYSA-N
Synonym 1-Chloro-4-ethoxybenzene
PubChem CID 69326
IUPAC Name 1-chloro-4-ethoxybenzene
SMILES CCOC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 214°C
Formula Weight 156.61
Percent Purity ≥99.0% (GC)
Chemical Name or Material 4-Chlorophenetole
EINECSNumber (3)-0845
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.