Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chlorophenoxy acetohydrazide

Catalog No. JRD08681G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD08681G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD08681G Supplier Sigma Aldrich Supplier No. JRD08681G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl bromopyruvate is an inhibitor of carbonic anhydrase.Methyl bromopyruvate was used in the synthesis of 2-(4-amino-4-carboxybutyl)thiazole-4-carboxylic acid and methyl-1-(bromomethyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-1-carboxylate.

Specifications

CAS 2381-75-1
Molecular Formula C8H9ClN2O2
Linear Formula C8H9ClN2O2
MDL Number MFCD00705185
Quantity 25 mL
Molecular Weight (g/mol) 200.62
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.