Learn More
Medchemexpress LLC 4-Chlorophenoxyacetic acid | 122-88-3 | 99.9% | 186.59 | 1 KG
SDP
Catalog No. 5000403346
Encompass_Preferred
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression, making it suitable for promoting fruit growth and controlling weeds. It is for research use only.
  • Mimics endogenous auxin
  • Regulates plant cell division, differentiation, and gene expression
  • Used to control weeds
  • Promotes fruit growth
Catalog No. 50-004-03346 Supplier Medchemexpress LLC Supplier No. HY196981KG
Add to Cart
Add to Cart
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression, making it suitable for promoting fruit growth and controlling weeds. It is for research use only.
  • Mimics endogenous auxin
  • Regulates plant cell division, differentiation, and gene expression
  • Used to control weeds
  • Promotes fruit growth