Learn More
Medchemexpress LLC 4-Chlorophenoxyacetic acid (4-CPA) | 122-88-3 | 99.9% | 186.59 | 100 G
SDP
Catalog No. 5000401799
Encompass_Preferred
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression. It is used to control weeds and promote fruit growth.
  • Promotes wild-type tomato fruit growth by upregulating LeExp2 and SAUR expression.
Catalog No. 50-004-01799 Supplier Medchemexpress LLC Supplier No. HY19698100G
Add to Cart
Add to Cart
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression. It is used to control weeds and promote fruit growth.
  • Promotes wild-type tomato fruit growth by upregulating LeExp2 and SAUR expression.