Learn More
Medchemexpress LLC 4-Chlorophenoxyacetic acid (4-CPA) | 122-88-3 | 99.9% | 186.59 | 25 G
SDP
Catalog No. 5000403293
Encompass_Preferred
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator that mimics endogenous auxin to regulate plant cell division, differentiation, and gene expression. It is primarily used to control weeds and promote fruit growth, also finding use as a laboratory chemical and in the manufacture of various substances.
  • Mimics endogenous auxin
  • Regulates plant cell division, differentiation, and gene expression
  • Promotes wild-type tomato fruit growth
  • Used to control weeds
  • Used to promote fruit growth
Catalog No. 50-004-03293 Supplier Medchemexpress LLC Supplier No. 25GHY19698
Add to Cart
Add to Cart
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator that mimics endogenous auxin to regulate plant cell division, differentiation, and gene expression. It is primarily used to control weeds and promote fruit growth, also finding use as a laboratory chemical and in the manufacture of various substances.
  • Mimics endogenous auxin
  • Regulates plant cell division, differentiation, and gene expression
  • Promotes wild-type tomato fruit growth
  • Used to control weeds
  • Used to promote fruit growth