Learn More
Medchemexpress LLC 4-Chlorophenoxyacetic acid (4-CPA) | 122-88-3 | 99.9% | 186.59 | 50 G
SDP
Catalog No. 5000402745
Encompass_Preferred
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression. It is used to control weeds and promote fruit growth.
  • Widely used herbicide and plant growth regulator.
  • Mimics endogenous auxin.
  • Regulates plant cell division, differentiation, and gene expression.
  • Used to control weeds and promote fruit growth.
Catalog No. 50-004-02745 Supplier Medchemexpress LLC Supplier No. HY1969850G
Add to Cart
Add to Cart
4-Chlorophenoxyacetic acid (4-CPA) is a widely used herbicide and plant growth regulator. It mimics endogenous auxin and regulates plant cell division, differentiation, and gene expression. It is used to control weeds and promote fruit growth.
  • Widely used herbicide and plant growth regulator.
  • Mimics endogenous auxin.
  • Regulates plant cell division, differentiation, and gene expression.
  • Used to control weeds and promote fruit growth.