Learn More
Medchemexpress LLC (4-chlorophenyl)-6-(2-methylindazol-5-yl)-2-(2,2,2-trifluoroethylamino)pyrido[4,3-d]pyrimidin-7-one | 2377491-54-6 | 99.5% | C23H16ClF3N6O | 10MG
SDP
Catalog No. 5000258359
Encompass_Preferred
AGI-43192 is a potent, orally active inhibitor of methionine adenosyltransferase 2A (MAT2A) with low-nanomolar cellular activity and reported antitumor effects in preclinical models. Supplied as a solid powder for biochemical and cellular research.
  • Potent MAT2A inhibition with low-nanomolar activity
  • Reduces S-adenosyl methionine (SAM) levels in MTAP-null cells
  • Demonstrated tumor growth inhibition in preclinical studies
  • High purity suitable for research applications
  • Available in multiple small research pack sizes
  • Stable under recommended storage conditions for long-term use
Catalog No. 50-002-58359 Supplier Medchemexpress LLC Supplier No. HY14577710MG
AGI-43192 is a potent, orally active inhibitor of methionine adenosyltransferase 2A (MAT2A) with low-nanomolar cellular activity and reported antitumor effects in preclinical models. Supplied as a solid powder for biochemical and cellular research.
  • Potent MAT2A inhibition with low-nanomolar activity
  • Reduces S-adenosyl methionine (SAM) levels in MTAP-null cells
  • Demonstrated tumor growth inhibition in preclinical studies
  • High purity suitable for research applications
  • Available in multiple small research pack sizes
  • Stable under recommended storage conditions for long-term use