Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl Chloroformate 98.0+%, TCI America™
SDP

Catalog No. C25455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25455G Supplier TCI America Supplier No. C25455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7693-45-0
Color Colorless
Boiling Point 102°C
Molecular Formula C7H4Cl2O2
Quantity 5 g
MDL Number MFCD00000638
UN Number 3277
Synonym 4-chlorophenyl chloroformate, 4-chlorophenylchloroformate, p-chlorophenyl chloroformate, carbonochloridic acid, 4-chlorophenyl ester, 4-chlorophenyl carbonochloridate, chloroformic acid 4-chlorophenyl ester, acmc-1biw7, 4-chloro-phenyl chloroformate, ksc493g4j, 4-chlorophenylcarbonochloridate
InChI Key RYWGPCLTVXMMHO-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OC(=O)Cl)Cl
IUPAC Name (4-chlorophenyl) carbonochloridate
Molecular Weight (g/mol) 191.007
PubChem CID 139059
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl Chloroformate
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.