Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4-Chlorophenyl)Triethoxysilane

Catalog No. 5979101G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-791-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-791-01G Supplier Sigma Aldrich Supplier No. 5979101G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4-Chlorophenyl)triethoxysilane can be used:To facilitate the connection between dipolar mixed monolayers andzinc oxide in photovoltaic devices.As a substrate in the Ni-catalyzed decarboxylative coupling with alkynyl carboxylic acids to yield substituted diarylalkynes.As a substrate in the Hiyama cross-coupling reactions with 3-iodoazetidines to yield substituted 3-arylazetidines.As an intermediate in the synthesis of tripod-shaped oligo(p-phenylene)s, which are used in the surface immobilization of gold and CdS quantum dots for sensor applications.

Specifications

CAS 429689-17-8
Melting Point 89°C to 93°C
Molecular Formula C19H19NO2
Refractive Index n20/D 1.474
Linear Formula C19H19NO2
MDL Number MFCD05664284
Quantity 1 g
Synonym Methyl (3S)-(+)-3-(methyl-1H-indol-3-yl)-3-phenylpropanoate
Molecular Weight (g/mol) 293.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.