Learn More
Medchemexpress LLC 4-Chlorophenylacetic acid | 1878-66-6 | 99.97% | 170.59 | 1000 G
SDP
Catalog No. 5000453178
Encompass_Preferred
4-Chlorophenylacetic acid is an analytical standard intended for research and analytical applications. It is a halogenated phenylacetic acid derivative, a potent aromatase inhibitor, and antagonizes estrogen signaling. This compound can provide carbon and energy for Pseudomonas sp. strain CBS3 and is effective against estrogen-induced mammary tumorigenesis.
  • Analytical standard for research and analytical applications.
  • Halogenated phenylacetic acid derivative.
  • Potent aromatase inhibitor.
  • Antagonizes estrogen signaling.
  • Provides carbon and energy for Pseudomonas sp. strain CBS3.
  • Effective against estrogen-induced mammary tumorigenesis.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Reference standard supplied assay.
Catalog No. 50-004-53178 Supplier Medchemexpress LLC Supplier No. HYW0100621000G
4-Chlorophenylacetic acid is an analytical standard intended for research and analytical applications. It is a halogenated phenylacetic acid derivative, a potent aromatase inhibitor, and antagonizes estrogen signaling. This compound can provide carbon and energy for Pseudomonas sp. strain CBS3 and is effective against estrogen-induced mammary tumorigenesis.
  • Analytical standard for research and analytical applications.
  • Halogenated phenylacetic acid derivative.
  • Potent aromatase inhibitor.
  • Antagonizes estrogen signaling.
  • Provides carbon and energy for Pseudomonas sp. strain CBS3.
  • Effective against estrogen-induced mammary tumorigenesis.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Reference standard supplied assay.