Learn More
Medchemexpress LLC 4-chlorophenylacetic acid | 1878-66-6 | 99.97% | 170.59 | 250 G
SDP
Catalog No. 5000453406
Encompass_Preferred
4-Chlorophenylacetic acid is a halogenated phenylacetic acid derivative. It acts as a potent aromatase inhibitor and antagonizes estrogen signaling. This compound can serve as a carbon and energy source for Pseudomonas sp. strain CBS3 and has shown effectiveness against estrogen-induced mammary tumorigenesis.
  • Halogenated phenylacetic acid derivative.
  • Potent aromatase inhibitor.
  • Antagonizes estrogen signaling.
  • Can provide carbon and energy for Pseudomonas sp. strain CBS3.
  • Effective against estrogen-induced mammary tumorigenesis.
Catalog No. 50-004-53406 Supplier Medchemexpress LLC Supplier No. 250GHYW010062
Add to Cart
Add to Cart
4-Chlorophenylacetic acid is a halogenated phenylacetic acid derivative. It acts as a potent aromatase inhibitor and antagonizes estrogen signaling. This compound can serve as a carbon and energy source for Pseudomonas sp. strain CBS3 and has shown effectiveness against estrogen-induced mammary tumorigenesis.
  • Halogenated phenylacetic acid derivative.
  • Potent aromatase inhibitor.
  • Antagonizes estrogen signaling.
  • Can provide carbon and energy for Pseudomonas sp. strain CBS3.
  • Effective against estrogen-induced mammary tumorigenesis.