Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylacetic acid, 98%

Catalog No. AC160720025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160720025 2.5 kg Plastic bottle
AC160720050 5 g Glass bottle
AC160721000 100 g Plastic bottle
AC160725000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC160720025 Supplier Thermo Scientific Chemicals Supplier No. 160720025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1878-66-6 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1

Specifications

Melting Point 103°C to 107°C
Color White to Cream
CAS Min % 97.5
CAS Max % 100.0
Quantity 2.5 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 97.5% min. (HPLC)
Linear Formula ClC6H4CH2CO2H
Beilstein 09,448
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,solubility 5% in chcl3: clear solution
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetic acid, 98%
Show More Show Less
EINECSNumber 217-521-6
RTECSNumber AG0590000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.