Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylacetonitrile, 98+%

Catalog No. AAA1529522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1529522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1529522 Supplier Thermo Scientific Chemicals Supplier No. A1529522

missing translation for 'validMainframeMsg'

CAS: 140-53-4 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-53-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001918
InChI Key IVYMIRMKXZAHRV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
PubChem CID 241582
ChEBI CHEBI:17346
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)Cl

Specifications

Melting Point 28°C to 31°C
Density 1.19
Boiling Point 141°C to 143°C (11 mmHg)
Flash Point 134°C (273°F)
Odor Pungent
Refractive Index 1.543
Quantity 100 g
UN Number UN3439
Beilstein 971171
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenylacetonitrile
Show More Show Less
Hazard Category H300-H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 205-418-9
RTECSNumber AL8250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.