Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-4-Chlorophenylglycine

Catalog No. 6917125GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-171-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 69-171-25GF Supplier Sigma Aldrich Supplier No. 6917125GF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-4-Chlorophenylglycine

Specifications

CAS 346457-41-8
Melting Point 226°C to 230°C
Molecular Formula C7H6BrNO2
Linear Formula C7H6BrNO2
MDL Number MFCD00579106
Quantity 25 g
Synonym (+/-)-alpha-Amino-4-chlorophenylacetic acid
Molecular Weight (g/mol) 216.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.